Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cetrimoniumbromid, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

0219502980 0219502990 0219502991
ICNA0219502980EA 48.2 EUR InStock
ICNA0219502980 ICNA0219502990 ICNA0219502991
Cetrimoniumbromid, weißes Pulver
Cetrimoniumbromid
A phase transfer catalyst ideal for the precipitation of DNA

  • HLB 10

Cetyltrimethylammonium bromide ia used as an cationic surfactant suitable for the isolation of high molecular weight DNA in plants and other organisms(by a rapid method) as well as plant DNA for use in PCR analysis. It has been used to precipitate nucleic acids. CTAB provides enhancement of Concanavalin A mediated agglutination. It has been used for the determination of protein molecular weights in electrophoretic systems and for determination of critical micelle concentration of detergents. CTAB has also been used as a titrant for potentiometric titration of perchlorate and as a phase-transfer catalyst in reduction of arenes and heterocyclic compounds.

Freely but slowly soluble in water; Freely soluble in methanol and ethanol; soluble in alcohol.

Achtung! Bitte beachten: Research Use Only (RUO).


Formel: CH₃(CH₂)₁₅N(Br)(CH₃)₃
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011772
CAS-Nummer: 57-09-0
UN: 3077
ADR: 9,III
Order Now
Spezifikation Testergebnisse
Identity Test Passes Passes
FTIR Conforms to Standard
Storage Store at Room Temperature (15 to 30 °C)
Aggregation Number 170
CMC 1 mM (20 - 25 °C)
pH 6,0 - 7,5 (0,1 M H2O, 0 °C) (Lit.)

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge