Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Calcein, orangenes Pulver Forschungsqualität

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0219508701 0219508705 0219508710
ICNA0219508701EA 35.1 EUR InStock
ICNA0219508701 ICNA0219508705 ICNA0219508710
Calcein, orangenes Pulver Forschungsqualität
Calcein
Calcein is used as an Indicator for the determination of calcium by titration with EDTA. Suitable for fluorometric determination of calcium.

Calcein does not fluoresce in strong alkaline solution, but an intense fluorescence appears in the presence of alkaline earths, especially calcium. The yellow green fluorescence disappears instantly with an addition of EDTA. Also in acidic solution, the yellow green fluorescence of Calcein disappears with an addition of copper, zinc and mercury.

Practically insoluble in water; easily soluble in alkaline solutions to give an orange red solution.


Formel: C₃₀H₂₆N₂O₁₃
Siedepunkt: 200 °C (Lit.)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00005049
CAS-Nummer: 1461-15-0
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge