Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Fluorescamin, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


0219518325 0219518380 0219518391
ICNA0219518325EA 40.7 EUR InStock
ICNA0219518325 ICNA0219518380 ICNA0219518391
Fluorescamin, weißes Pulver
Fluorescamine
Fluorescamine is a non-fluorescent reagent that reacts readily with primary amino groups in amino acids and peptides to form highly fluorescent derivatives, whereas the reagent and its degradation products are non-fluorescent. No significant fluorescence with ammonia. Low background due to hydrolysis.
an plasma and urine and sulfonamides in honey by HPLC with fluorescence detection.

  • Fluorescence: Emmax: 475 nm; Exc.: 390 nm in 0.5 M borate buffer, pH 8.5, after derivatization with L-leucine

Fluorescamine is a reagent for fluorescent assay of amino acids, peptides, proteins and other primary amines. It is also used for fluorometric assays of some proteolytic enzymes. It effectively blocks newly generated amino termini in protein sequence analyses by the Beckman automated sequencer. This product has been used in labeling casein so that it can be used as a substrate for measuring protease activity.

Fluorescamine is used in many sensitive detection methods, eg. characterization of poly-L-lysine (pLL)/DNA complexes post-modified with a multivalent hydrophilic polymer, spectrofluorimetric analysis of procaine, detection and quantitation of residual aminopenicillins by HPLC after fluorescamine derivation, determination of lisinopril in hum

Soluble in CH₂Cl₂, trichloromethane, chloroform, dimethysulphoxide; slightly soluble in acetone (50 mg/ml), ethanol and water.


Formel: C₁₇H₁₀O₄
Melting Pt: 148…158 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00005928
CAS-Nummer: 38183-12-9
Order Now
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge