Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Propidiumiodid, powder_dark red

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

0219545810 0219545825 0219545880 0219545883
ICNA0219545810EA 48.3 EUR InStock
ICNA0219545810 ICNA0219545825 ICNA0219545880 ICNA0219545883
Propidiumiodid, powder_dark red
Propidiumiodid
Propidium iodide is a fuorescent stain for nucleic acids.

Cell membrane integrity excludes propidium iodide from staining viable and apoptotic cells. Propidium iodide may be used in flow cytometry to evaluate cell viability when used with other dyes that stain viable cells or cells that are early in the apoptosis process.

Propidium iodide (PI) binds to DNA by intercalating between the bases with little or no sequence preference and with a stoichiometry of one dye per 4 to 5 base pairs of DNA. PI also binds to RNA, necessitating treatment with nucleases to distinguish between RNA and DNA staining. Once the dye is bound to nucleic acids, its fluorescence is enhanced 20- to 30-fold, the fluorescence excitation maximum is shifted ~30 to 40 nm to the red and the fluorescence emission maximum is shifted ~15 nm to the blue. Although its molar absorptivity (extinction coefficient) is relatively low, PI exhibits a sufficiently large Stokes shift to allow simultaneous detection of nuclear DNA and fluorescein-labeled antibodies, provided the proper optical filters are used. Propidium iodide is suitable for fluorescence microscopy, confocal laser scanning microscopy, flow cytometry and fluorometry.

Soluble at 1 mg/ml in water.


Formel: C₂₇H₃₄I₂N₄
MW: 668.4 g/mol
MDL Number: MFCD00011921
CAS-Nummer: 25535-16-4
Order Now
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard
UV/VIS: l max (water) 493 ± 4 nm

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge