Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Trimethoprim, weißes Pulver

Lieferant: MP BIOMEDICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

0219552701 0219552705 0219552725 0219552780
ICNA0219552701EA 47.7 EUR InStock
ICNA0219552701 ICNA0219552705 ICNA0219552725 ICNA0219552780
Trimethoprim, weißes Pulver
Trimethoprim
Trimethoprim is an antibacterial agent; a specific inhibitor of formylation; a dihydrofolate reductase inhibitor.
Trimethoprim is primarily used as an antibacterial agent. It has dihydrofolate reductase inhibitor activity with selectivity for the prokaryote enzyme. It is used to treat urinary tract infections, mild acute prostatitis, recurrent cystitis, asymptomatic bacteriuria during pregnancy and respiratory tract infections.
The combination of trimethoprim and sulfamethoxazole interferes with the cellular metabolism of folic acid in the bacterial cell by blocking the biosynthesis of nucleotides. Trimethoprim binds to dihydrofolate reductase and inhibits the reduction of dihydrofolic acid (DHF) to tetrahydrofolic acid (THF). THF is an essential precursor in the thymidine synthesis pathway and interference with this pathway inhibits bacterial DNA synthesis.

  • Presentation: White Powder
  • Soluble in a hot 1:1 solution of ethanol:chloroform (50 mg/mL - clear, colorless to faint yellow solution); soluble in chloroform (15 mg/mL - clear, colorless solution), propylene glycol (25 mg/mL), methanol (12 mg/mL); very slightly soluble in water (0.4 mg/mL).
  • +4 °C

Trimethoprim is an antibacterial agent; a specific inhibitor of formylation; a dihydrofolate reductase inhibitor.

Trimethoprim is primarily used as an antibacterial agent. It has dihydrofolate reductase inhibitor activity with selectivity for the prokaryote enzyme. It is used to treat urinary tract infections, mild acute prostatitis, recurrent cystitis, asymptomatic bacteriuria during pregnancy and respiratory tract infections.


Formel: C₁₄H₁₈N₄O₃
MW: 290.32 g/mol
Melting Pt: 196…206 °C
Storage Temperature: Kühlschrank
MDL Number: MFCD00036761
CAS-Nummer: 738-70-5
Order Now
Spezifikation Testergebnisse
Identity Test Passes
FTIR Conforms to Standard

 

PRODUKT

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge