Vitamin Diet Fortification Powder, MP Biomedicals
Lieferant: MP Biomedicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0290465401
ICNA0290465401EA 280 EUR InStock
ICNA0290465401
Vitamin Diet Fortification Powder, MP Biomedicals
Tierbedarf Tiernahrung
Vitamin diet fortification mixture formulation provides sufficient vitamin levels to prevent deficiencies in all animal species, giving precise control of vitamin levels.
The ingredients of the vitamin diet fortification mixture are triturated in dextrose (1 kg is sufficient for 100 lbs of diet).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge