Vitamin mixture, AIN-76-A, MP Biomedicals
Lieferant: MP Biomedicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0296009801
ICNA0296009801EA 163 EUR InStock
ICNA0296009801
Vitamin mixture, AIN-76-A, MP Biomedicals
Tierbedarf Tiernahrung
Basic component of animal diet especially for maintaining mice and rats colonies in the research laboratory.
Please see product datasheet for complete diet composition.
Lieferumfang: In powdered form.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge