Tris-Glycine (TG) buffer,, MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
04816200
ICNA04816200EA 83.9 EUR InStock
ICNA04816200
Tris-Glycine (TG) buffer,, MP Biomedicals
Puffer Elektrophorese-Puffer Laufpuffer
Tris-glycine buffer is used for western blotting and gel electrophoresis.
Tris-glycine buffer is used to make a Tris-glycine-methanol transfer buffer, which is the most common protein transfer buffer for wet blot transfers. The methanol prevents the gel from swelling during the transfer and enhances the protein binding to nitrocellulose.
Conveniently preweighed and premixed, the buffers are packaged in foil packets. Simply place the contents of one packet in a 4 litrer flask, and add deionised water.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge