Vector, pUC 18, MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
04821226
ICNA04821226EA 214 EUR InStock
ICNA04821226
Vector, pUC 18, MP Biomedicals
Vektoren, Plasmide und Bibliotheken
pUC 18 is a small vector of approximately 2,7 Kb containing the Pvu II/EcoR I fragment of pBR322 and the lacZ gene multiple cloning site from M13mp18.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge