Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IgG Horse Antibody

Lieferant: MP Biomedicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

08641441
ICNA08641441EA 207 EUR InStock
ICNA08641441
Anti-IgG Horse Antibody
Antikörper
The immunoglobulin was prepared from horse serum using current chromatographic techniques. A quantitative
protein analysis has been applied to determine quality of the product.

  • Supplied in Tris or phosphate buffered saline
  • Can be reconstitute with 0,135 M Sodium chloride

It is useful for structural and biological studies, as a reference standard, or as starting material for the preparation of an immunogen.

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Horse
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge