Freund's complete adjuvant, MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
08642851
ICNA08642851EA 169 EUR InStock
ICNA08642851
Freund's complete adjuvant, MP Biomedicals
Protein- Expression Proteinexpressions-Reagenzien
Freund's complete adjuvant is made by a modification of Freund's technique, in which 25 mg of Mycobacterium is suspended in a mixture of 7,5 ml Arlacel A and 42,5 ml paraffin oil.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge