Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Synaptophysin IgG1 Mouse Monoclonal Antibody [clone: SY38]

Lieferant: MP Biomedicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

08697301
ICNA08697301EA 794 EUR InStock
ICNA08697301
Anti-Synaptophysin IgG1 Mouse Monoclonal Antibody [clone: SY38]
Antikörper
This antibody identifies synaptophysin, a 38 kDa membrane glycoprotein. It may be a valuable aid in the examination of various neuropathological disorders, especially those associated with altered synapse patterns and defects of neurotransmitter sections. It may also possess potential for studying diseases such as Alzheimer's disease, alcoholic fetoembryopathy, and senile dementia.

  • Specificity: It is an excellent marker for several neuroendocrine, neuronal and adrenal tumors, and reacts with synaptophysin, a transmembrane glycoprotein of presynaptic vesicles
  • Dilution range: 1:50 for Immunohistochemical application
  • It can reconstitute with 1,0 ml of distilled water

The antibody is in PBS buffer, pH 7,4, with 0,5% BSA, and 0,09% sodium azide.

Type: Primary
Antigen: Synaptophysin
Clonality: Monoclonal
Clone: SY38
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat, Rat PC-12 cell line

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge