RPMI 1640 (1X), Cell Culture Media, MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
091629149 091646454
ICNA091629149EA 69.3 EUR InStock
ICNA091629149 ICNA091646454
RPMI 1640 (1X), Cell Culture Media, MP Biomedicals
Zellkulturmedien
RPMI has successfully been used for the cultivation of normal human and neoplastic leukocytes. It is now a popular general purpose medium when properly supplemented. As a modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent.
  • Originally developed by Moore and his colleagues at Roswell Park Memorial Institute (RPMI)
  • Widely used in fusion protocols and in the growth of hybrid cells
  • Used with a serum supplement, to support several cell lines in the absence of serum
It was based on the RPMI 1630 line of media which utilised a bicarbonate buffering system and alterations in the amounts of amino acids and vitamins.
The Dutch Modification of RPMI 1640 is also available through MP, contains 6400 mg/L sodium chloride instead of 6000 mg/L, 1000 mg/L sodium bicarbonate instead of 2000 mg/L, and 20 mM HEPES.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge