Cryopreservation media, Cellvation, MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0920300M2
ICNA0920300M2EA 332 EUR InStock
ICNA0920300M2
Cryopreservation media, Cellvation, MP Biomedicals
Zelleinfriermedien
Cellvation is a cryopreservation medium designed for optimal cell recovery and convenience.
  • Optimal cell recovery and convenience
  • No DMSO or serum
  • Animal component free
  • Preserve stem cells, hepatocytes, and tissue samples.
Cellvation does not contain DMSO or serum, and is scientifically engineered for serum-free cultures, as well as being ideally suited for all types of serum. Cellvation cryopreservation media is animal-component-free and has been shown to preserve the potential of stem cells for differentiation. In addition, the medium is recommended for the preservation of hepatocytes, tissue samples, and a wide variety of cell types and established cell lines.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge