DNA purification kits, Geneclean® kit, MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
111001200
ICNA111001200EA 261 EUR InStock
ICNA111001200
DNA purification kits, Geneclean® kit, MP Biomedicals
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
The GeneClean® Kit is designed to purify fragments of DNA from 200 bp to 20 kb from solutions and agarose gels.
Applications include desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearised from uncut vector, and isolation of PCR products away from genomic DNA.
The kit simplifies the process of purifying DNA into three easy steps: Bind, wash and elute. Ethanol precipitation is never required and purified DNA is immediately ready for a wide variety of downstream applications.
Zubehörinformationen: Geneclean® Glassmilk™ is a specially prepared aqueous suspension of proprietary silica matrix that allows flexibility of reaction size and volume. Geneclean® Glassmilk™ binds to radioactive isotope-labeled ssDNA or dsDNA purified from agarose resulting in purified DNA suitable for further use.
Lieferumfang: Supplied with Geneclean® Glassmilk™, which is also available separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge