Transformation kit, EZ-Yeast™, MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
112100200
ICNA112100200EA 108 EUR InStock
ICNA112100200
Transformation kit, EZ-Yeast™, MP Biomedicals
Nukleinsäure-Reagenzien Transformations-Reagenzien
The EZ-Yeast™ Transformation Kit is designed for high throughput transformation with efficiencies up to 10³ transformants per μg plasmid DNA. It is ideal for cases where a large number of transformations must be performed, but only a few transformants are needed. Perfect for a quick analysis of potentially positive two-hybrid colonies, it does not require fully competent cells prior to transformation.
Lieferumfang: Kit contains EZ-Transformation Solution and carrier DNA.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge