Yeast transformation kit, Alkali-Cation™, MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
112200200
ICNA112200200EA 495 EUR InStock
ICNA112200200
Yeast transformation kit, Alkali-Cation™, MP Biomedicals
Nukleinsäure-Reagenzien Transformations-Reagenzien
Alkali-Cation™ Yeast Transformation Kit provides an efficient method for yeast transformation and has been successfully used to transform linear DNA into Pichia pastoris for protein expression studies.
  • Easy to use – can be completed in less than 90 minutes
  • High transformation efficiency routinely greater than 10⁴ transformants per µg of DNA
  • Scalability allows high efficiency transformation needed in two hybrid screens
The kit provides all of the reagents required to perform an easy and quick non electrical method for transforming yeast with linear or plasmid DNA. Host cells are made competent by treatment with a lithium/caesium acetate mixture.
Lieferumfang: Kit includes pH 7,5 TE; lithium/acesium acetate; carrier DNA, 10 mg/ml; sterile histamine solution; TE/cation MIXX; and sterile solution SOS.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge