Ethidium bromide Greenbag™ disposal kit, MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
112350200
ICNA112350200EA 254 EUR InStock
ICNA112350200
Ethidium bromide Greenbag™ disposal kit, MP Biomedicals
Elektrophorese-Reagenzien Reagenzien zur Entfernung von Farbstoffen
The EtBr Greenbag™ Disposal Kit allows rapid and trouble-free concentration of ethdium bromide from large volumes of solutions into a small 'teabag' containing activated carbon. The Greenbag™ is then conveniently disposed of along with other solid hazardous wastes.
One kit has the capacity to remove 500 mg of ethidium bromide from solutions (10 mg EtBr/bag), enough to satisfy the requirements of most laboratories for a year.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge