116961100
116540438
116961001
116961050
116961500
116978025
116978050
116985050
116985025
116985100
116978005
116540439
ICNA116961100EA
357
EUR
InStock
ICNA116961100
ICNA116540438
ICNA116961001
ICNA116961050
ICNA116961500
ICNA116978025
ICNA116978050
116985050.
116985025.
116985100.
116978005.
116540439.
Lysing matrix Z, MP Biomedicals
Protein-Extraktion
Proteinextraktions-Reagenzien
Lysing Matrix Z is used primarily for lysis of animal and plant tissue, tougher stems, roots and seeds, and whole insects. Each impact-resistant tube contains yttria-stabilised zirconium oxide beads.
- Impact-resistant tube
- Ideal for cracking of very tough cell walls
- Lower shear rate minimises degradation
- Free from DNase and RNase
Matrix Z is an alternative to the very aggressive Matrix A because it's lower shear rate minimises degradation in applications such as RNA and protein isolation from very tough tissue.