Taq DNA polymerase, MP Biomedicals
Lieferant: MP BIOMEDICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
11EPTQD025 11EPTQD325 11EPTQC200 11EPTQD925 11EPTQA105 11EPTQA925 11EPTQA100 11EPTQA025 11EPTQA325 11EPTQC060 11EPTQL100
ICNA11EPTQD025EA 163 EUR InStock
ICNA11EPTQD025 ICNA11EPTQD325 ICNA11EPTQC200 ICNA11EPTQD925 ICNA11EPTQA105 ICNA11EPTQA925 ICNA11EPTQA100 ICNA11EPTQA025 ICNA11EPTQA325 ICNA11EPTQC060 ICNA11EPTQL100
Taq DNA polymerase, MP Biomedicals
Nukleinsäure-Reagenzien Enzyme und Kits für Endpunkt-PCR
Taq DNA Polymerase is a specialised thermostable enzyme isolated from the thermophilic bacterium Thermus aquaticus. The recombinant form of this enzyme is expressed in E. coli. It is highly processive, recombinant polymerase which catalyses the addition of mononucleotide units to the 3’-OH end of a primer chain. It remains functional even after prolonged incubation at 95 °C. This enzyme possesses 5’-3’ exonuclease activiy.
Taq DNA Polymerase is used for the DNA polymerase chain reaction (PCR) in order to amplify DNA sequences. Other applications include RT-PCR, DNA sequencing and DNA labeling.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge