Chlorite ion chromatography (IC) standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ICS006-100ML ICS006-500ML
ICS006-100MLEA 112 EUR InStock
ICS006-100ML ICS006-500ML
Chlorite ion chromatography (IC) standard, Supelco®
Standards Anionische Standards Chlorit-Standards
Chlorite IC standard may be used in the determination of disinfection by-product, chlorite in water samples using ion chromatography technique (IC)
Zertifizierungen: his certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge