Handheld digital microscopes, special light, stroboscopic
Lieferant: Dino-Lite / IDCP B.V.
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AM3713TB AM3715TB
IDCPAM3713TBEA 439 EUR InStock
IDCPAM3713TB IDCPAM3715TB
Handheld digital microscopes, special light, stroboscopic
Mikroskope
Portable microscopes with stroboscopic technology to capture fast moving objects.
  • Perfect pictures thanks to reduced motion blur, even under higher magnification
  • Stroboscopic light mode can be switched on and off
  • High speed USB 2.0 output
  • Edge series model with high-quality optics provides a very sharp, bright and natural colour image with very low aberration and vignetting
  • AM3715TB features exchangeable caps and an external trigger to synchronise picture capturing
  • Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
Ideal for use in monitoring production lines in a manufacturing environment, or observing subjects such as a tadpole in a laboratory.
Lieferumfang: Supplied with carrying pouch and software (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)).
Achtung! Bitte beachten: Please note: For optimal results the environmental lighting should be lower than normal and the microscope should be closer to the object being viewed.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge