Digital Microscope
Lieferant: Dino-Lite / IDCP B.V.
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AM8917MZT
IDCPAM8917MZTEA 1600 EUR InStock
IDCPAM8917MZT
Digital Microscope
Mikroskope
Pushing the limits of handheld digital microscopy, the 8-megapixel AM8917MZT is a high-resolution digital microscope capable of revealing the minute details of the target. The microscope also delivers outstanding colour fidelity, enhancing the imaging experience with detailed and natural-looking results. Ready to perform a wide range of tasks, AM8917MZT offers a polarizer as well as DPQ, EDOF, EDR, AMR, and eFLC.
  • High optical resolution
  • Enhanced flexible LED control (eFLC)
  • Automatic magnification reading (AMR)
  • Robust metal housing
  • Extended depth of field (EDOF)
  • Extended dynamic range (EDR)
  • Adjustable polariser
2 years warranty.
Verpackung: N3C-C (close cap) N3C-D (diffuser cap) N3C-D2 (opal diffuser cap) N3C-E (extended open cap) N3C-L (long cap) N3C-O (open cap) N3C-S (sidelight cap).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge