Handheld digital microscopes, Dino-Lite Medical, TrichoScope
Lieferant: Dino-Lite / IDCP B.V.
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MEDL3H MEDL7HM
630-2608EA 329 EUR InStock
630-2608 IDCPMEDL7HM
Handheld digital microscopes, Dino-Lite Medical, TrichoScope
Mikroskope
These handheld microscopes are ideal tools for studying the scalp, hair shaft and the hair itself in detail (trichoscopy). Images made with the TrichoScope can help the specialists to recognise skin or hair problems and diseases.
  • Built-in polariser filter (some models only) minimises the glaring effect of the scalp/hair
  • High speed USB 2.0 connection
  • Powered through USB connection: Do not need batteries
  • Polariser models: Built-in 8 LEDs can be enabled or disabled as desired; an external light source is unnecessary
  • Robust composite/plastic or metal housing
Zertifizierungen: Medical Device Directive 93/42/CEE modified 2007/47/EC.
Lieferumfang: Supplied with DinoCapture (Windows) or DinoXcope software (MacOS) and carry pouch.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge