Microscope clamp, gooseneck, Dino-Lite RK-02
Lieferant: Dino-Lite / IDCP B.V.
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
RK-02
IDCPRK-02EA 149 EUR InStock
IDCPRK-02
Microscope clamp, gooseneck, Dino-Lite RK-02
Klemmen Klemmen für den allgemeinen Gebrauch
The RK-02 is a durable gooseneck clamp designed for flexible and versatile usage with any Dino-Lite digital microscope.
  • C-clamp fits to virtually any surface up to 7 cm (~3 inch) thick
  • Can be mounted horizontally or vertically
With this sturdy stand, users can easily position the Dino-Lite by bending the 60 cm long arm and adjusting the rotating head. The base of the RK-02 can also be rotated providing even more freedom and it can be fixed with the quick release fastener for use on uneven surfaces.
Compatible with all Dino-Lite models, except the high magnification models.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge