Digital Microscope
Lieferant: Dino-Lite / IDCP B.V.
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WF7915MZT
IDCPWF7915MZTEA 929.4 EUR InStock
IDCPWF7915MZT
Digital Microscope
Mikroskope
With the use of the latest, cutting-edge optics, a brand new 5 megapixel sensor and several special features, it has a marvel of technology and the best choice for the high-demanding professional. It offers superb image quality and color reproduction in a robust, compact and appealing housing.
  • Wireless
  • Extended depth of field (EDOF)
  • Extended dynamic range (EDR)
  • Flexible LED control (FLC)
  • Automatic magnification reading (AMR)
With the Extended Dynamic Range (EDR) feature, the details of darker or brighter areas within the object can be revealed by stacking images at different exposure levels. The Extended Depth of Field (EDOF) feature automatically stacks images at different focus level to improve the depth of field on rough or uneven surfaces. With the built-in Automatic Magnification Reading (AMR), measurements can be performed easily and quickly. Because of the built-in polarization filter this model is ideal when working with shiny or reflective objects such as metal, plastic, glass, jewelry, electronics, etc.
Provides 2 years warranty.
Verpackung: Microscope, carry pouch, software CD, calibration target, user manual, N3C-O- open cap, N3C-C closed cap, N3C-D- diffuser cap, N3C-E extension cap, N3C-L- Long cap, N3C-S- side light cap.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge