Closed cap, Dino-Lite
Lieferant: Dino-Lite / IDCP B.V.
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ZT-Z-CC1
IDCPZT-Z-CC1EA 7.95 EUR InStock
IDCPZT-Z-CC1
Closed cap, Dino-Lite
Mikroskope
Closed cap addition for the polarising AM series (ZT models). Designed to help protect the lens from dust and more.
This snap-on cover fits the AM413ZT, AM4013MZT and AM7013MZT model. Not designed for use with the AD series or AM models without polarisation.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge