Magnetrührer RCT 5 digital
Lieferant: IKA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
20002704
IKAA20002704EA 1201 EUR InStock
IKAA20002704
Magnetrührer RCT 5 digital
Rührer Magnetrührer mit Heizplatte
The RCT 5 digital magnetic hotplate stirrer offers significantly higher performance with heat output of approximately 800 W. This hotplate stirrer is ideal for a wide range of heating and stirring tasks.
  • Automatic reverse rotation with intermittend mode
  • Multifunctional menu
  • Viscosity trend measurement
  • Digital LED display shows actual and set speed setting
  • Timer and sensor in medium detection (via PT1000 sensor)
  • Can be used at 5 to 40 °C (80% relative humidity), in an incubator or cold room
Whether determining the direction of rotation, timer functions or sequence programming - the stirring process can be programmed individually from beginning to end. In this way, the reaction runs reliably and safely even without being monitored.
The white coated ceramic plate has an angular shape to maximise space.
Zertifizierungen: IP protection class according to DIN EN 60529: IP 42.
Lieferumfang: Wird mit USB-Kabel, PT1000,60 Temperaturfühler und zwei Magnetstäbchen geliefert.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge