Electronic overhead stirrer, NANOSTAR 7.5 digital
Lieferant: IKA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
25004356
IKAA25004356EA 986 EUR InStock
IKAA25004356
Electronic overhead stirrer, NANOSTAR 7.5 digital
Rührer Rührwerke
Small and space saving overhead stirerr with great perfomance.
  • Guarantees a constant torque over the entire rpm range from 50 to 2000 min⁻¹
  • Easy to operate using a stable rotary knob
  • Display clearly shows the rotational speed
  • Regular software updates can be undertaken rapidly and simply via the USB interface
The NANOSTAR 7.5 digital overhead stirrer is 30% smaller than the MICROSTAR 7.5 digital and therefore even more space-saving. However, its performance remains the same. NANOSTAR 7.5 digital convinces with perfect basic functionalities, its compact design and weighing just 0,8 kg means it is easy to handle.
Zertifizierungen: IP protection class according to DIN EN 60529: IP 54
Bestellinformation: Supplied without accessories, a wide range of accessories are available to order separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge