Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KRT Mouse Monoclonal Antibody [clone: LU5]

Lieferant: IMMUNOLOGIC

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

ILM1026-C01 ILM1026-C05 ILM1026-C1 ILM1026-R10 ILM1026-R25 S/ILM1026-R2
ILM1026-C01EA 242 EUR InStock
ILM1026-C01 ILM1026-C05 ILM1026-C1 ILM1026-R10 ILM1026-R25 VWRKS/ILM1026-R2
Anti-KRT Mouse Monoclonal Antibody [clone: LU5]
Antikörper
Lu-5 reacts with an epitope which is common to all cytokeratins respectively with a cytokeratin-associated protein. The antigen recognized by anti-cytokeratin pan is present in all types of epithelia. It recognizes most subtypes of cytokeratins in cryofixed and paraffin sections and is ideally suited as a first order pan-epithelial marker. The epitope recognized by Lu-5 is a formalin-resistant marker of great value in tumour diagnosis, located on the surface of cytokeratin filaments. The epitope is present in most cytokeratin polypeptides of both the acidic (type I) and basic (type II) subfamily but does not occur in other cytoskeletal proteins. The epithelial specificity and the broad tissue and species crossreactivity provide an excellent probe for the differential diagnosis of epithelial versus mesenchymal tumours, large cell lymphomas and neural tumours.

  • Control: adenocarcinoma, skin and squamous carcinoma
  • Dilution Range: 1:100-1:500

Pretreatment: Protease K treatment for 10 minutes at 37 °C is required for immunohistochemical staining on formalin-fixed, paraffin embedded tissue sections.

Type: Primary
Antigen: KRT
Clonality: Monoclonal
Clone: LU5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge