Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IgG Goat Polyclonal Antibody (Biotin (NHS-LC))

Lieferant: ImmunoReagents

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

IR1436
IMMRIR1436EA 148 EUR InStock
IMMRIR1436
Anti-IgG Goat Polyclonal Antibody (Biotin (NHS-LC))
Antikörper
ImmunoReagents’ Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualized, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Zertifizierungen: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.

Bestellinformation: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Donkey

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge