Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: ImmunoReagents

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

IR1900
IMMRIR1900EA 222 EUR InStock
IMMRIR1900
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
ImmunoReagents Alkaline Phosphatase (AP) antibody conjugates are used in ELISA, Immunohistochemistry (IHC) and Western blotting techniques. AP is a 140 kDa enzyme that changes color in the presence of PNPP or BCIP/NBT substrates. These conjugates have a linear reaction rate thus allowing the researcher to increase sensitivity by prolonging substrate incubation times.

  • Origin: US

ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Zertifizierungen: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.

Bestellinformation: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge