Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: IMMUNOREAGENTS
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
PRODUKT
Erweitert / Kompakt
Secondary Antibody Goat anti-rat IgG (H&L) - Affinity Pure, min x w/Hu or Mu IgG (highly absorbed against Mu IgG), ALP Conjµgate, H ost: Goat, Format: Liq, Label: ALP, Applications: WB, ELISA 1 * 1 mg IMMRIR2247 1 * 1 mg
Hersteller-Artikelnummer:: IR2247
Lokale Artikelnummer:: IMMRIR2247
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder