Pump tubing, Tygon® MHSL 2001
Lieferant: ISMATEC PART OF COLE PARMER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SC0818
ISMCSC0818EA 0 EUR InStock
ISMCSC0818
Pump tubing, Tygon® MHSL 2001
Schläuche Transferschlauch
The transparent, plasticiser-free tubing with superior pump-life. Eespecially designed for MEK and other aggressive solvents.
  • Plasticizer and oil-free
  • Smooth inner-bore
  • Low sorption maintains fluid and tube integrity
  • Does not impart anything into the pumping medium
  • No release of hazardous materials when properly incinerated
  • Complies with FDA standards
Temperature resistant from −73 to +57 °C
Hardness (Shore durometer A): 69
Density: 0,88
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge