Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Heating tapes, IT-SiS10

Lieferant: ISOPAD

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

115096-000 162874-000
632-4801EA 211.8 EUR InStock
632-4801 ISOP162874-000
Heating tapes, IT-SiS10
Heizgeräte
Isopad IT-SiS10 is a preterminated, flexible, and waterproof silicone insulated heating tape. Four silicone insulated heating conductors are embedded in a protective overbraid made of cupro-nickel. The temperature depends on the power and application.

  • Maximum operating temperature: 200 °C
  • Width×Thickness: 11×5 mm (±10%)
  • Maximum power per meter 100 W/m

It is non hazardous and used in ordinary areas. It is used for temperature modification of pipes and structures in industries and laboratories.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge