PCR plates, 96-well, fully skirted, exo-frame
Lieferant: iST Scientific Ltd
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
IST-601-096GCT
ISTSIST-601-096GCTEA 419 EUR InStock
ISTSIST-601-096GCT
PCR plates, 96-well, fully skirted, exo-frame
Mikrotiterplatten PCR-Platten
Fully skirted 96-well PCR plate with clear PP wells and grey PC frame.
  • Free of DNA, DNase, RNase, and PCR inhibitors
  • Low binding PP wells
  • Alpha numberic labeling
  • Manufactured under the most modern clean room conditions
  • Automation friendly
Rigid two-component design eliminates warping and distortion during PCR.
Verpackung: Packaged 10 plates per sleeve, 50 plates per box.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge