J60040.09
J60040.18
J60040.09EA
29.5
EUR
InStock
J60040.09
J60040.18
N-Lauroylsarkosin Natriumsalz 95%
N-Lauroylsarkosin Natriumsalz
Purity: 95%
CAS No.: 137-16-6
Molecular Formula: C15H29NNaO3
Formula Weight: 293.39
Physical Form: Crystalline powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 44-47°
Beilstein Reference: 5322974
Merck Reference: 14,4368
MDL No.: MFCD00042728
Used in the cell lysis step of RNA purification. Anionic detergent