Leu-Enkephalin
Lieferant: ALFA AESAR
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
J61408.MB
J61408.MBEA 97.5 EUR InStock
J61408.MB
Leu-Enkephalin
Proteine und Peptide
CAS No.: 81678-16-2 Molecular Formula: C28H37N5O7 Formula Weight: 555.70 Storage Temperature: -30°C to -10°C Physical Form: Lyophilized powder Appearance: White MDL No.: MFCD00076406
Applications: Endogenous opioid neurotransmitter that is an agonist at mu- and delta-opioid receptors
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge