J61457.06
J61457.09
J61457.14
J61457.06EA
92.5
EUR
InStock
J61457.06
J61457.09
J61457.14
Furosemid ≥97%
Furosemide
Purity: 97+%
CAS No.: 54-31-9
Molecular Formula: C12H11ClN2O5S
Formula Weight: 330.74
Physical Form: Crystalline powder
Appearance: Off-white
Melting Point: 206-209°
Solubility: Soluble in acetone, DMF or methanol. Slightly soluble in water
MDL No.: MFCD00010549
Applications: Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter