Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Neomycin trisulphate salt hydrate

Lieferant: ALFA AESAR

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

J61499.14 J61499.22
J61499.14EA 48 EUR InStock
J61499.14 J61499.22
Neomycin trisulphate salt hydrate
Neomycin trisulphate salt hydrate
Neomycin sulfate is an aminoglycoside antibiotic and calcium channel protein inhibitor. Neomycin sulfate also binds to prokaryotic ribosomes inhibiting translation and is effective against gram-positive and gram-negative bacteria.

Neomycin sulfate inhibits Phospholipase C via binding to inositol phospholipids. It also inhibits phosphatidylcholine-PLD activity and induces Ca2+ mobilization and PLA2 activation in human platelets. Neomycin sulfate inhibits DNase I induced DNA degradation.

An antibiotic. Acts as an inhibitor of protein biosynthesis


Formel: C₂₃H₄₆N₆O₁₃·₃H₂SO₄·xH₂O
Storage Temperature: Raumtemperatur
MDL Number: MFCD00150829
CAS-Nummer: 1405-10-3
EINECS: 215-773-1
Merck Index: 14,6454
Order Now
Spezifikation Testergebnisse
Potency 600 µg/mg min.
Loss on Drying 8.0% max.

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge