J61772.06
J61772.06EA
73.8
EUR
InStock
J61772.06
Sulindac
Sulindac
CAS No.: 38194-50-2
Molecular Formula: C20H17FO3S
Formula Weight: 356.41
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Yellow
Melting Point: 182-185°
UN No.: UN2811
MDL No.: MFCD00599589
Applications: A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells