J61871.MD
J61871.03
J61871.MDEA
247
EUR
InStock
J61871.MD
J61871.03
D(-)-Milchsäure
D(-)-Milchsäure
CAS No.: 10326-41-7
Molecular Formula: C3H6O3
Formula Weight: 90.08
Storage Temperature: 2°C to 8°C
Physical Form: Low melting solid
Appearance: White to off-white
Solubility: Soluble in water (50mg/ml)
Beilstein Reference: 1720254
Merck Reference: 14,5335
Optical Rotation: -0.4° (c=8 in water)
MDL No.: MFCD00068313
Useful chiral synthon, building block for depsipeptides