Schiff-Reagenz
Lieferant: ALFA AESAR
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Achtung
J62171.AP
J62171.APEA 109 EUR InStock
J62171.AP
Schiff-Reagenz
Schiff-Reagenz
Used for detection of glycoproteins in polyacrylamide gels.
Intended for use in Sigma Periodic Acid Staining Procedure, 395. Kit is intended for use with blood, bone marrow, tissue touch preparations or routine tissue sections. It is used for staining glycoproteins in polyacrylamide gels. Schiff's reagent is also sometimes used to dye organic tissues. It is been widely used for detection of a certain functional group in chemical solutions.
Used for detection of glycoproteins in polyacrylamide gels
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge