Nefazodone hydrochloride ≥98%
Lieferant: ALFA AESAR
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
J62793.03
J62793.03EA 115 EUR InStock
J62793.03
Nefazodone hydrochloride ≥98%
Nefazodone hydrochloride
Purity: 98+% CAS No.: 82752-99-6 Molecular Formula: C25H32ClN5O2.HCl Formula Weight: 506.47 Physical Form: Powder Appearance: White Melting Point: 186-187° MDL No.: MFCD00935760
Applications: An antidepressant acts by modifying serotonin transmission. Mixed 5-HT2A serotonin receptor antagonist/serotonin uptake inhibitor
Formel: C₂₅H₃₂ClN₅O₂·HCl
Melting Pt: 186…187 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00935760
CAS-Nummer: 82752-99-6
Merck Index: 14,6438
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge