J62811.06
J62811.14
J62811.22
J62811.06EA
63.7
EUR
InStock
J62811.06
J62811.14
J62811.22
Nifedipin 98%
Nifedipin
Purity: 98%
CAS No.: 21829-25-4
Molecular Formula: C17H18N2O6
Formula Weight: 346.33
Physical Form: Powder
Appearance: Off-white to yellow
Melting Point: 171-175°
MDL No.: MFCD00057326
Applications: L-type calcium channel blocker; induces apoptosis in human glioblastoma cells