J62873.03
J62873.06
J62873.14
J62873.03EA
43
EUR
InStock
J62873.03
J62873.06
J62873.14
L-Methioninsulfoxid
L-Methioninsulfoxid
CAS No.: 3226-65-1
Molecular Formula: C5H11NO3S
Formula Weight: 165.21
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 4129939
MDL No.: MFCD00063093