J63254.06
J63254.06EA
60.2
EUR
InStock
J63254.06
Fendiline hydrochloride
Fendiline hydrochloride
CAS No.: 13636-18-5
Molecular Formula: C23H25N.HCl
Formula Weight: 351.92
Physical Form: Powder
Appearance: White
Melting Point: 204-205°
MDL No.: MFCD00079301
Applications: A calcium channel blocker