J63295.MC
J63295.03
J63295.MCEA
56.2
EUR
InStock
J63295.MC
J63295.03
Nicergolin ≥95%
Nicergolin
CAS No.: 27848-84-6
Molecular Formula: C24H26BrN3O3
Formula Weight: 484.39
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00869626
Applications: Cerebral and peripheral vasodilator