J63472.MA
J63472.MB
J63472.MF
J63472.MAEA
144
EUR
InStock
J63472.MA
J63472.MB
J63472.MF
6-Cyan-7-nitrochinoxalin-2,3-dion 98%
6-Cyan-7-nitrochinoxalin-2,3-dion
Purity: 99+%
CAS No.: 115066-14-3
Molecular Formula: C9H4N4O4
Formula Weight: 232.16
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 300°
Solubility: Soluble in DMSO to 100mM
Beilstein Reference: 5821072
MDL No.: MFCD00069232
Potent, selective kainate/quisqualate (non-NMDA) receptor antagonist