J63772.MC
J63772.03
J63772.06
J63772.MCEA
77.6
EUR
InStock
J63772.MC
J63772.03
J63772.06
Sotalol hydrochloride 98%
Sotalol hydrochloride
Purity: 98%
CAS No.: 959-24-0
Molecular Formula: C12H20N2O3S.HCl
Formula Weight: 308.82
Physical Form: Crystalline powder
Appearance: White to off-white
MDL No.: MFCD00242937
Applications: Potent beta adrenergic antagonist